missing translation for 'onlineSavingsMsg'
Learn More

beta-Ketoglutaric Acid, TRC

Product Code. 30403499
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403499 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403499 Supplier Toronto Research Chemicals Supplier No. TRCK191205100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material beta-Ketoglutaric Acid
CAS 542-05-2
Molecular Formula C5 H6 O5
Quantity 100 g
Synonym 3-oxo-Glutaric Acid, β-oxo-Glutaric Acid, 1,3-Dicarboxyacetone, 3-Ketoglutaric Acid, 3-Oxoglutaric Acid, 3-Oxopentanedioic Acid, Acetone-1,3-dicarboxylic Acid, s-Acetonedicarboxylic Acid, β-Oxoglutaric Acid
InChI Formula InChI=1S/C5H6O5/c6-3(1-4(7)8)2-5(9)10/h1-2H2,(H,7,8)(H,9,10)
SMILES OC(=O)CC(=O)CC(=O)O
IUPAC Name 3-oxopentanedioic acid
Molecular Weight (g/mol) 146.098
Formula Weight 146.0215
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.