missing translation for 'onlineSavingsMsg'
Learn More

Beta-Isothipendyl Hydrochloride Salt, TRC

Product Code. 30392120
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392120 100 mg
30400950 10 mg
30389648 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392120 Supplier Toronto Research Chemicals Supplier No. TRCI902751100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132962-42-6
Molecular Formula C16 H19 N3 S . Cl H
Molecular Weight (g/mol) 321.8681
Synonym 10H-Pyrido[3,2-b][1,4]benzothiazine-10-ethanamine, N,N,β-trimethyl-, hydrochloride (1:1), 10H-Pyrido[3,2-b][1,4]benzothiazine, 10-(2-dimethylamino-1-methylethyl)-, monohydrochloride (6CI), N,N,β-Trimethyl-10H-pyrido[3,2-b][1,4]benzothiazine-10-ethanamine Hydrochloride, β-Isothipendyl Hydrochloride
IUPAC Name N,N-dimethyl-2-pyrido[3,2-b][1,4]benzothiazin-10-ylpropan-1-amine;hydrochloride
SMILES Cl.CC(CN(C)C)N1c2ccccc2Sc3cccnc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H19N3S.ClH/c1-12(11-18(2)3)19-13-7-4-5-8-14(13)20-15-9-6-10-17-16(15)19;/h4-10,12H,11H2,1-3H3;1H
Formula Weight 321.1066
Chemical Name or Material beta-Isothipendyl Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.