missing translation for 'onlineSavingsMsg'
Learn More

beta-Ionone, 96%

Product Code. 11455413
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455413 500 g 500g
11445413 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455413 Supplier Thermo Scientific Alfa Aesar Supplier No. A13400.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 79-77-6 | C13H20O | 192.302 g/mol

It is an important fragrance chemical used in perfumery. The combination of α-ionone and β-ionone is characteristic of the scent of violets and used with other components in flavoring to recreate their scent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important fragrance chemical used in perfumery. The combination of α-ionone and β-ionone is characteristic of the scent of violets and used with other components in flavoring to recreate their scent.

Solubility
Soluble in water (0.11 mg/mI at 20°C).

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-77-6
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.302
MDL Number MFCD00001549
InChI Key PSQYTAPXSHCGMF-BQYQJAHWSA-N
Synonym beta-ionone, trans-beta-ionone, e-beta-ionone, 4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, .beta.-ionone, 3e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, 4-2,6,6-trimethyl-1-cyclohexenyl-3-buten-2-one, unii-a7nrr1hlh6, e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, beta-e-ionone
PubChem CID 638014
ChEBI CHEBI:32325
IUPAC Name (E)-4-(2,6,6-trimethylcyclohexen-1-yl)but-3-en-2-one
SMILES CC1=C(C(CCC1)(C)C)C=CC(=O)C

Specifications

Melting Point -49°C
Density 0.943
Boiling Point 126°C to 128°C (12 mmHg)
Flash Point 112°C (233°F)
Refractive Index 1.52
Quantity 500 g
Beilstein 1909544
Merck Index 14,5056
Solubility Information Soluble in water (0.11mg/mI at 20°C).
Formula Weight 192.3
Percent Purity 96%
Chemical Name or Material beta-Ionone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.