missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Beta III Tubulin Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142939
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize isoform 1 (NP_006077.2) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.
The betaIII-tubulin isoform is present dominantly in cells of neuronal origin and it is one of the earliest markers of neuronal differentiation. It is regarded as a specific probe for the cells of neuronal origin as well as for the tumours originating from these cells. The betaIII-tubulin is most abundant in cells of neuronal origin, but was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues.
Specifications
| Beta III Tubulin | |
| Polyclonal | |
| Unconjugated | |
| TUBB3 | |
| 3200002H15Rik; beta 3 tubulin; beta tubulin 3; beta-4; betaIII-tubulin; CDCBM; CDCBM1; CFEOM3; CFEOM3A; class III beta-tubulin; FEOM3; I79_000194; LOC100341581; M(beta)3; M(beta)6; mc1R; melanocortin receptor 1; Neuron specific beta III Tubulin; neuron-specific class III beta-tubulin; TUBB; Tubb3; TUBB4; tubulin beta; tubulin beta 3 class III; Tubulin beta-3; tubulin beta-3 chain; Tubulin beta-4; Tubulin beta-4 chain; tubulin beta-III; tubulin, beta 3 class III; tubulin, beta, 3; tubulin, beta, 4; Unknown (protein for MGC:134115) | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10381 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q13509 | |
| TUBB3 | |
| Peptide with sequence C-DPSGNYVGDSD. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction