missing translation for 'onlineSavingsMsg'
Learn More

beta-III Tubulin Mouse anti-Human, Clone: TUBB3/3731, Novus Biologicals™

Product Code. 18309781 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18309781 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18309781 Supplier Novus Biologicals Supplier No. NBP30738520UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-III Tubulin
Applications Western Blot, Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone TUBB3/3731
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Alias beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
Host Species Mouse
Immunogen A synthetic peptide (aa 437-450) of human beta-III Tubulin protein (TUBB3); coupled to KLH.
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 10381
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.