missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-III Tubulin Mouse anti-Human, Clone: TUBB3/3731, Novus Biologicals™
Mouse Monoclonal Antibody
£401.00
Specifications
| Antigen | beta-III Tubulin |
|---|---|
| Clone | TUBB3/3731 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
Description
beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
| beta-III Tubulin | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 10381 | |
| IgG1 κ | |
| Protein A or G purified |
| TUBB3/3731 | |
| Western Blot, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| A synthetic peptide (aa 437-450) of human beta-III Tubulin protein (TUBB3); coupled to KLH. | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title