missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-III Tubulin Antibody (CL5814), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61431-25ul
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifikationer
| beta-III Tubulin | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
| TUBB3 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: YEDDEEESEAQGPK | |
| 25 μL | |
| Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers, Neuroscience | |
| 10381 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL5814 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:50000 - 1:100000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:50000 - 1:100000 | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specificity of human beta-III Tubulin antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering