missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-III Tubulin Antibody (CL5813) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44243
This item is not returnable.
View return policy
Description
beta-III Tubulin Monoclonal antibody specifically detects beta-III Tubulin in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence,Immunohistochemistry
Specifications
| beta-III Tubulin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 μg/mL, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:50000 - 1:100000 | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| This antibody was generated using a synthetic peptide of Q13509, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience, Phospho Specific | |
| 10381 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence, Immunohistochemistry | |
| CL5813 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction