missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta II Tubulin B Antibody (5B2), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-74037UV
This item is not returnable.
View return policy
Description
beta II Tubulin B Monoclonal specifically detects beta II Tubulin B in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| beta II Tubulin B | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TUBB2B | |
| Recombinant human beta-Tubulin (1-445aa) purified from E. coli | |
| 100 μL | |
| Signal Transduction | |
| 347733 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 5B2 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| bA506K6.1, class II beta-tubulin isotype, DKFZp566F223, FLJ98847, MGC8685, tubulin beta-2B chain, tubulin, beta 2B, tubulin, beta polypeptide paralog | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human beta-Tubulin mAb, clone 5B2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human beta-Tubulin protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion