missing translation for 'onlineSavingsMsg'
Learn More

beta II Tubulin Antibody (OTI3B7), Novus Biologicals Biologicals™

Product Code. 30306496 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30306496 100 μL 100µL
30306647 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30306496 Supplier Novus Biologicals Supplier No. NBP344865100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta II Tubulin Monoclonal antibody specifically detects beta II Tubulin in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta II Tubulin
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone OTI3B7
Conjugate Unconjugated
Dilution Western Blot 1:4000, Immunocytochemistry/ Immunofluorescence 1:100
Formulation PBS (pH 7.3) 1% BSA, 50% glycerol
Host Species Mouse
Immunogen Full length human recombinant protein of human TUBB2A(NP_001060) produced in HEK293T cell.
Purification Method >95%
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.