missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta Ig-h3/TGFBI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£280.00 - £458.00
Specifications
| Antigen | Beta Ig-h3/TGFBI |
|---|---|
| Dilution | Simple Western 1:25, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18431851
|
Novus Biologicals
NBP1-88606-25ul |
25 μL |
£280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18711074
|
Novus Biologicals
NBP1-88606 |
0.1 mL |
£458.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Beta Ig-h3/TGFBI Polyclonal specifically detects Beta Ig-h3/TGFBI in Human samples. It is validated for Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Beta Ig-h3/TGFBI | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Beta ig-h3, BIGH3EBMD, CDB1, CDG2, CDGG1transforming growth factor, beta-induced, 68kD, CSD, CSD1, CSD2, CSD3, Kerato-epithelin, LCD1, RGD-CAP, RGD-containing collagen-associated protein, transforming growth factor, beta-induced, 68kDa, transforming growth factor-beta-induced protein ig-h3 | |
| TGFBI | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Simple Western 1:25, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Cytokine Research, Immunology, Vision | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 7045 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DHHATNGVVHLIDKVISTITNNIQQIIEIEDTFETLRAAVAASGLNTMLEGNGQYTLLAPTNEAFEKIPSETLNRILGDPEALRDLLNNHILKSAMCAEAIVAGLSVETLEGTTLEVGCSGDMLTINGKAIISNKD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title