missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta Ig-h3/TGFBI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20613
This item is not returnable.
View return policy
Description
Beta Ig-h3/TGFBI Polyclonal specifically detects Beta Ig-h3/TGFBI in Human, Chicken samples. It is validated for Western Blot.
Specifications
| Beta Ig-h3/TGFBI | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| Beta ig-h3, BIGH3EBMD, CDB1, CDG2, CDGG1transforming growth factor, beta-induced, 68kD, CSD, CSD1, CSD2, CSD3, Kerato-epithelin, LCD1, RGD-CAP, RGD-containing collagen-associated protein, transforming growth factor, beta-induced, 68kDa, transforming growth factor-beta-induced protein ig-h3 | |
| Rabbit | |
| 75 kDa | |
| 0.1 mL | |
| Angiogenesis, Cytokine Research, Immunology, Vision | |
| 7045 | |
| Human, Chicken | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q15582 | |
| TGFBI | |
| Recombinant protein encompassing a sequence within the center region of human TGFBI. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Chicken. Species reactivity based on sequence identity: Mouse (85%), Rat (85%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction