missing translation for 'onlineSavingsMsg'
Learn More

Beta Ig-h3/TGFBI Antibody (320305) [FITC], Novus Biologicals™

Product Code. 30732386 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732386 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732386 Supplier Novus Biologicals Supplier No. FAB2559F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Beta Ig-h3/TGFBI Monoclonal antibody specifically detects Beta Ig-h3/TGFBI in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beta Ig-h3/TGFBI
Applications Western Blot
Classification Monoclonal
Clone 320305
Conjugate FITC
Formulation PBS
Gene Alias Beta ig-h3, BIGH3EBMD, CDB1, CDG2, CDGG1transforming growth factor, beta-induced, 68kD, CSD, CSD1, CSD2, CSD3, Kerato-epithelin, LCD1, RGD-CAP, RGD-containing collagen-associated protein, transforming growth factor, beta-induced, 68kDa, transforming growth factor-beta-induced protein ig-h3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse βIG-H3, Gly24-His683, Accession # P82198
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cytokine Research, Immunology, Vision
Primary or Secondary Primary
Gene ID (Entrez) 7045
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.