missing translation for 'onlineSavingsMsg'
Learn More

beta I Tubulin Rabbit anti-Human, Mouse, Rat, Bovine, Canine, Monkey, Clone: 1C3J0, Novus Biologicals™

Product Code. 18332002
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18332002 100 μg 100µL
18311316 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18332002 Supplier Bio-Techne Supplier No. NBP315928100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

beta I Tubulin Monoclonal antibody specifically detects beta I Tubulin in Human, Mouse, Rat, Bovine, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta I Tubulin
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1C3J0
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias beta tubulin 1, class VI, dJ543J19.4, tubulin beta-1 chain, tubulin, beta 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 352-451 of human beta I Tubulin (Q9H4B7).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 81027
Target Species Human, Mouse, Rat, Bovine, Canine, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.