missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
beta I Tubulin Monoclonal antibody specifically detects beta I Tubulin in Human, Mouse, Rat, Bovine, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
| Antigen | beta I Tubulin |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | SR1866 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:3000-1:10000, Flow Cytometry 1:100, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:200 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | beta tubulin 1, class VI, dJ543J19.4, tubulin beta-1 chain, tubulin, beta 1 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human beta I Tubulin (Uniprot #: Q9H4B7) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?