missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta I Tubulin Antibody (OTI4A3), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72225AF594
This item is not returnable.
View return policy
Description
beta I Tubulin Monoclonal specifically detects beta I Tubulin in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| beta I Tubulin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TUBB1 | |
| Full length recombinant protein of human beta I Tubulin (NP_110400) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI4A3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| beta tubulin 1, class VI, dJ543J19.4, tubulin beta-1 chain, tubulin, beta 1 | |
| Mouse | |
| Affinity Purified | |
| Cytoskeleton Markers | |
| 81027.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction