missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta I Tubulin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00081027-D01P
This item is not returnable.
View return policy
Description
beta I Tubulin Polyclonal antibody specifically detects beta I Tubulin in Human, Mouse samples. It is validated for Western Blot
Specifications
| beta I Tubulin | |
| Polyclonal | |
| PBS (pH 7.4) | |
| beta tubulin 1, class VI, dJ543J19.4, tubulin beta-1 chain, tubulin, beta 1 | |
| TUBB1 (NP_110400.1, 1 a.a. - 451 a.a.) full-length human protein. MREIVHIQIGQCGNQIGAKFWEMIGEEHGIDLAGSDRGASALQLERISVYYNEAYGRKYVPRAVLVDLEPGTMDSIRSSKLGALFQPDSFVHGNSGAGNNWAKGHYTEGAELIENVLEVVRHESESCDCLQGFQIVHSLGGGTGSGMGTLLMNKIREEYPDRIMNSFSVMPSPKVSDTVVEPYNAVLSIHQLIENADACFCIDNEALYDICFRTLKLTTPTYGDLNHLVSLTMSGITTSLRFPGQLNADLRKLAVNMVPFPRLHFFMPGFAPLTAQGSQQYRALSVAELTQQMFDARNTMAACDLRRGRYLTVACIFRGKMSTKEVDQQLLSVQTRNSSCFVEWIPNNVKVAVCDIPPRGLSMAATFIGNNTAIQEIFNRVSEHFSAMFKRKAFVHWYTSEGMDINEFGEAENNIHDLVSEYQQFQDAKAVLEEDEEVTEEAEMEPEDKGH | |
| 0.1 mg | |
| Cell Biology, Cellular Markers | |
| 81027 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| NP_110400.1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction