missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta I Tubulin Antibody (2A1A9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£163.00 - £442.00
Specifications
| Antigen | beta I Tubulin |
|---|---|
| Clone | 2A1A9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18218880
|
Novus Biologicals
NBP2-52577 |
0.1 mg |
£442.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650077
|
Novus Biologicals
NBP2-52577-0.025mg |
0.025 mg |
£163.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta I Tubulin Monoclonal specifically detects beta I Tubulin in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| beta I Tubulin | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cytoskeleton Markers | |
| PBS with 0.05% Sodium Azide | |
| 81027 | |
| Purified recombinant fragment of human TUBB1 (AA: 33-166) expressed in E. Coli. | |
| Primary | |
| 50.3 kDa |
| 2A1A9 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Monkey | |
| beta tubulin 1, class VI, dJ543J19.4, tubulin beta-1 chain, tubulin, beta 1 | |
| TUBB1 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title