missing translation for 'onlineSavingsMsg'
Learn More

Beta Hydroxysteroid Dehydrogenase Antibody (1E8), Novus Biologicals™

Product Code. 18329837 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329837 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329837 Supplier Novus Biologicals Supplier No. H00003284M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Beta Hydroxysteroid Dehydrogenase Monoclonal antibody specifically detects Beta Hydroxysteroid Dehydrogenase in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beta Hydroxysteroid Dehydrogenase
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1E8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000189
Gene Alias 3 beta-HSD type II, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 2, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type II, 3-beta-HSD II, delta 5-delta 4-isomerase type II, HSD3B, HSDB, HSDB3B, hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2,3-beta-hydroxy-Delta(5)-steroid dehydrogenase, progesterone reductase, SDR11E2, short chain dehydrogenase/reductase family 11E, member 2,3-beta-hydroxy-5-ene steroid dehydrogenase
Host Species Mouse
Immunogen HSD3B2 (NP_000189.1, 33 a.a. ~ 122 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ALDKAFRPELREEFSKLQNRTKLTVLEGDILDEPFLKRACQDVSVVIHTACIIDVFGVTHRESIMNVNVKGTQLLLEACVQASVPVFIYT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3284
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.