missing translation for 'onlineSavingsMsg'
Learn More

beta-Hydroxyisovaleric Acid, TRC

Product Code. 30132731
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132731 1 g 1g
30134385 50 g 50g
30008187 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132731 Supplier Toronto Research Chemicals Supplier No. TRCH9436051G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-08-1
Molecular Formula C5 H10 O3
Molecular Weight (g/mol) 118.13
Synonym Butanoic acid, 3-hydroxy-3-methyl-, Butyric acid, 3-hydroxy-3-methyl- (6CI,7CI,8CI), 3-Hydroxy-3-methylbutanoic acid, 3-Hydroxy-3-methylbutyric acid, 3-Hydroxyisovaleric acid, 3-Methyl-3-hydroxybutyric acid, β-Hydroxy-β-methylbutyric acid, β-Hydroxyisovaleric acid
IUPAC Name 3-hydroxy-3-methylbutanoic acid
SMILES CC(C)(O)CC(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H10O3/c1-5(2,8)3-4(6)7/h8H,3H2,1-2H3,(H,6,7)
Formula Weight 118.063
Chemical Name or Material beta-Hydroxyisovaleric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.