missing translation for 'onlineSavingsMsg'
Learn More

beta-Hydroxy-GABA Antibody, Novus Biologicals™

Product Code. 18396532
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396532 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396532 Supplier Bio-Techne Supplier No. NBP313407

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

beta-Hydroxy-GABA Polyclonal antibody specifically detects beta-Hydroxy-GABA in Rat samples. It is validated for ELISA, Immunofluorescence, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-Hydroxy-GABA
Applications ELISA, Immunofluorescence, Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias 4-Amino-3-hydroxybutanoic acid, beta-hydroxy-gamma-aminobutyric acid, Bussamina, Buxamina, Buxamine, GABOB, Gamibetal, gamma-Amino-beta-hydroxybutyric acid
Host Species Rabbit
Immunogen The immunogen used to generate this antibody corresponds to beta-Hydroxy-GABA
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.