missing translation for 'onlineSavingsMsg'
Learn More

Beta-Hexaacetylbromorutinose, TRC

Product Code. 30390474
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390474 5 mg 5mg
30404798 10 mg 10mg
30406303 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390474 Supplier Toronto Research Chemicals Supplier No. TRCH2652055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material beta-Hexaacetylbromorutinose
Analyte or Component Names beta-Hexaacetylbromorutinose
Shipping Condition Room Temperature
Molecular Formula C26H37BrO13
InChI Formula InChI=1S/C26H37BrO13/c1-11-8-20(24(38-15(5)31)25(39-16(6)32)21(11)35-12(2)28)34-10-18-9-19(27)23(37-14(4)30)26(40-17(7)33)22(18)36-13(3)29/h11,18-26H,8-10H2,1-7H3/t11-,18-,19+,20-,21+,22-,23+,24+,25-,26+/m1/s1
Solution Type Neat
SMILES C[C@H]1[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H](OC(C)=O)[C@H](OC[C@H]2C[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]2OC(C)=O)C1
IUPAC Name (1R,2S,3R,4S,6R)-4-bromo-6-((((1R,2S,3R,4S,5R)-2,3,4-triacetoxy-5-methylcyclohexyl)oxy)methyl)cyclohexane-1,2,3-triyl triacetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 637.47
Quantity 5 mg
Formula Weight 637.14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.