missing translation for 'onlineSavingsMsg'
Learn More

Beta-Geranyl Acetate, TRC

Product Code. 30009682
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009682 1 g 1g
30008723 10 g 10g
30011035 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009682 Supplier Toronto Research Chemicals Supplier No. TRCG3672101G
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-87-3
Molecular Formula C12 H20 O2
Molecular Weight (g/mol) 196.29
Synonym 2,6-Octadien-1-ol, 3,7-dimethyl-, 1-acetate, (2E)-, 2,6-Octadien-1-ol, 3,7-dimethyl-, acetate, (2E)- (9CI), 2,6-Octadien-1-ol, 3,7-dimethyl-, acetate, (E)- (8CI), Geraniol acetate (6CI), (E)-3,7-Dimethyl-2,6-octadien-1-ol acetate, (E)-3,7-Dimethyl-2,6-octadienyl acetate, 3,7-Dimethyl-2,6-octadien-1-ol acetate, Acetic acid (2E)-3,7-dimethyl-2,6-octadienyl ester, Acetic acid geraniol ester, Bay pine (oyster) oil, Geranyl acetate, Geranyl ethanoate, NSC 2584, trans-1-Acetoxy-3,7-dimethyl-2,6-octadiene, trans-3,7-Dimethyl-2,6-octadien-1-yl acetate, trans-Geranyl acetate, β-Geranyl acetate
IUPAC Name [(2E)-3,7-dimethylocta-2,6-dienyl] acetate
SMILES CC(=CCC\C(=C\COC(=O)C)\C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H20O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,8H,5,7,9H2,1-4H3/b11-8+
Formula Weight 196.1463
Chemical Name or Material Geranyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.