missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ beta-GalNAc-PEG4-Azide

Product Code. 18715676
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18715676 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18715676 Supplier Tocris Bioscience™ Supplier No. FAB10988S100UG
  • ONLINE EXCLUSIVE PRICE
    £438.00
    £413.70 / 25mg
    Save £24.30
    5% Off
 Restricted Item
Estimated Shipment: 16-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Asialoglycoprotein receptor ligand with PEG4 linker and azide group for onward chemistry

β-GalNAc-PEG4-Azide is a functionalized asialoglycoprotein receptor (ASGPR) ligand for lysosomal targeting chimera (LYTAC) research and development; incorporates a single ASGPR ligand with a PEG4 linker and azide group reactive handle ready for conjugation. It can be used as a building block for multivalent compounds to enhance ASGPR binding. Upon binding to ASGPR, β-GalNAc conjugates are efficiently internalized via ASGPR-mediated endocytosis. β-GalNAc conjugation can be employed as a strategy to effectively deliver cargo such as RNA or Cas9 complexes in a cell-specific manner to hepatocytes. Can be used to generate LYTACs, or labeled with dye for tissue imaging.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl 2-(acetylamino)-2-deoxy-β-D-galactopyranoside
CAS 879004-92-9
Quantity 25 mg
Target ASGPR1
Molecular Formula C16H30N4O9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.