missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Beta-Galactosidase (E. coli)

Product Code. 16262250
Click to view available options
Quantity:
1000 U
Unit Size:
1000 units
This item is not returnable. View return policy

Product Code. 16262250

Brand: Abnova™ P5270.1000Units

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Formulation In 1.6M ammonium sulfate
Molecular Weight (g/mol) 540kDa
Name Beta-Galactosidase (E. coli)
Preparation Method Native protein chromatographically purified from E. coli
Quantity 1000 U
Storage Requirements Store at 4°C. This product is stable for at least 3 months.
Regulatory Status RUO
Common Name BETA-GALACTOSIDASE
Assays Beta Galactosidase Assay
Biological Activity ≥300 units per mg protein. One Unit hydrolyzes one micromole of o-nitrophenyl-β-D-galactopyranoside per minute at 25°C, pH 7.5.
Species E. coli
Protein Tag None
Expression System Native protein chromatographically purified from E. coli
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.