missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Galactosidase, Armenian Hamster anti-E. coli, Clone: 13R4, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP2-81033-0.025MG
This item is not returnable.
View return policy
Description
beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, Flow Cytometry, Control.
Specifications
| beta Galactosidase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Control | |
| EC 3.2.1.23; b-gal | |
| beta Galactosidase. | |
| 0.025 mg | |
| Primary | |
| E. coli | |
| Purified |
| Western Blot, Flow Cytometry | |
| 13R4 | |
| Unconjugated | |
| PBS | |
| Armenian Hamster | |
| Protein A purified | |
| RUO | |
| 13R4 recognizes wild-type E. coli beta-galactosidase. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction