missing translation for 'onlineSavingsMsg'
Learn More

beta Galactosidase, Armenian Hamster anti-E. coli, Clone: 13R4, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Brand:  Novus Biologicals NBP2-81033-0.2MG

 View more versions of this product

Product Code. 18205719

  • £510.00 / 0.20mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, Flow Cytometry, Control.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

beta Galactosidase
Monoclonal
1 mg/mL
Western Blot, Flow Cytometry, Control
EC 3.2.1.23; b-gal
beta Galactosidase.
0.2 mg
Primary
E. coli
Purified
Western Blot, Flow Cytometry
13R4
Unconjugated
PBS
Armenian Hamster
Protein A purified
RUO
13R4 recognizes wild-type E. coli beta-galactosidase.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG λ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.