missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Galactosidase Antibody (BG-02) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-334
This item is not returnable.
View return policy
Description
beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| beta Galactosidase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| EC 3.2.1.23; b-gal | |
| beta-Galactosidase purified from E. coli. | |
| 0.1 mg | |
| Beta Galactosidase Epitope Tags, Epitope Tags, Fusion Proteins | |
| The antibody BG-02 recognizes a non-catalytic centre epitope of beta-galactosidase from E. coli. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| BG-02 | |
| Unconjugated | |
| Q56987 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| E. coli | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction