missing translation for 'onlineSavingsMsg'
Learn More

beta Galactosidase Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18412246 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412246 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412246 Supplier Novus Biologicals Supplier No. NBP176322AF488

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chicken Polyclonal Antibody

beta Galactosidase Polyclonal antibody specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Galactosidase
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias EC 3.2.1.23; b-gal
Host Species Chicken
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of E. coli beta-gal. The immunogen is located within amino acids 20 - 70 of beta-Galactosidase. Amino Acid Squence: WRNSEEARTDRPSQQLR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags, Fusion Proteins
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.