missing translation for 'onlineSavingsMsg'
Learn More

beta Galactosidase Antibody (13R4), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18406356 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406356 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406356 Supplier Novus Biologicals Supplier No. NBP281033MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Armenian Hamster Monoclonal Antibody

beta Galactosidase Monoclonal antibody specifically detects beta Galactosidase in E. coli samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Galactosidase
Applications Flow Cytometry
Classification Monoclonal
Clone 13R4
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias EC 3.2.1.23; b-gal
Host Species Armenian Hamster
Immunogen beta Galactosidase.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.