missing translation for 'onlineSavingsMsg'
Learn More

beta Galactosidase Antibody (13R4), FITC, Novus Biologicals™

Product Code. 18353290 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353290 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353290 Supplier Novus Biologicals Supplier No. NBP281032F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Galactosidase
Applications Flow Cytometry
Classification Monoclonal
Clone 13R4
Conjugate FITC
Dilution Flow Cytometry
Formulation PBS
Gene Alias EC 3.2.1.23; b-gal
Host Species Human
Immunogen beta Galactosidase.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity 13R4 recognizes wild-type E. coli beta-galactosidase.
Target Species E. coli
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.