missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Galactosidase Antibody (13R4), Allophycocyanin, Novus Biologicals™
Shop All Bio Techne ProductsDescription
beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | beta Galactosidase |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 13R4 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | EC 3.2.1.23; b-gal |
| Host Species | Human |
| Immunogen | beta Galactosidase. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?