missing translation for 'onlineSavingsMsg'
Learn More

beta-Estradiol, 99% (dry wt.), ca 3% water

Product Code. 11386656
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386656 25 g 25g
11366656 1 g 1g
11376656 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11386656 Supplier Thermo Scientific Alfa Aesar Supplier No. L03801.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 50-28-2 | C18H24O2 | 272.388 g/mol

β-Estradiol is the major estrogen secreted by the premenopausal ovary. It acts as an estrogenic hormone receptor (ER) agonist and activator of PI 3-kinase. Further, it is used in the treatment of infertility in women, prostate cancer and breast cancer. It is also employed as an agent of stimulating female puberty induction, as in the treatment of delayed puberty caused by hypogonadism or Turner's syndrome. In addition to this, it is used as part of the hormone replacement therapy for transgender women.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
β-Estradiol is the major estrogen secreted by the premenopausal ovary. It acts as an estrogenic hormone receptor (ER) agonist and activator of PI 3-kinase. Further, it is used in the treatment of infertility in women, prostate cancer and breast cancer. It is also employed as an agent of stimulating female puberty induction, as in the treatment of delayed puberty caused by hypogonadism or Turner′s syndrome. In addition to this, it is used as part of the hormone replacement therapy for transgender women.

Solubility
Soluble in dimethyl sulfoxide, ethanol , water, phosphate buffer saline, dimethyl formamide, acetone, dioxane and alkali hydroxides. Slightly soluble in vegetable oils.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-28-2
Molecular Formula C18H24O2
Molecular Weight (g/mol) 272.388
MDL Number MFCD00003693
InChI Key VOXZDWNPVJITMN-ZBRFXRBCSA-N
Synonym estradiol, beta-estradiol, 17beta-estradiol, oestradiol, dihydrofolliculin, estrace, ovocyclin, progynon, dihydrotheelin, dihydroxyestrin
PubChem CID 5757
ChEBI CHEBI:16469
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O

Specifications

Melting Point 175°C to 178°C
Quantity 25 g
Beilstein 1914275
Merck Index 14,3703
Solubility Information Soluble in dimethyl sulfoxide,ethanol ,water,phosphate buffer saline,dimethyl formamide,acetone,dioxane and alkali hydroxides. Slightly soluble in vegetable oils.
Optical Rotation +79° (c=1 in Dioxane)
Formula Weight 272.39
Percent Purity 99%
Assay (dry wt.), ≈3% water
Chemical Name or Material beta-Estradiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.