missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Beta Endorphin Recombinant Rabbit Monoclonal Antibody (20H4L2)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ 701295

Product Code. 15337114

  • £442.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with mouse, rat, non-human primate and rabbit based on sequence homology. Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Beta endorphin is an opioid neuropeptide that is produced by neurons of the pituitary gland and released into the circulatory system, or produced by the hypothalamus, and released into the brain and spinal cord through nerve fibers. The precursor polypeptide for beta endorphin is pro-opiomelanocortin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Beta Endorphin
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide
P01189
POMC
Peptide corresponding to amino acids 242-267 of human Beta Endorphin.
100 μg
Primary
Human
Antibody
IgG
Western Blot
20H4L2
Unconjugated
POMC
ACTH; Adrenocorticotropic hormone; adrenocorticotropin; alpha-melanocyte-stimulating hormone; alpha-MSH; Beta-endorphin; beta-LPH; beta-melanocyte-stimulating hormone; beta-MSH; CLIP; Corticotropin; Corticotropin-like intermediary peptide; corticotropin-lipotropin; Gamma-LPH; gamma-MSH; Lipotropin beta; lipotropin gamma; LPH; Melanocyte-stimulating hormone alpha; Melanocyte-stimulating hormone beta; Melanotropin alpha; melanotropin beta; Melanotropin gamma; met-enkephalin; MSH; NPP; opiomelanocortin prepropeptide; OTTHUMP00000119991; OTTHUMP00000200964; POC; POMC; Potential peptide; Precursor of MSH; pro-ACTH-endorphin; proopiomelanocortin; pro-opiomelanocortin; proopiomelanocortin preproprotein
Rabbit
Protein A
RUO
5443
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.