missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-endorphin Antibody (B31.15) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-414
This item is not returnable.
View return policy
Description
Beta-endorphin Monoclonal specifically detects Beta-endorphin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Beta-endorphin | |
| Monoclonal | |
| 1.0 mg/mL | |
| Immunohistochemistry 10 to 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL, Immunohistochemistry-Frozen 1:200 | |
| ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody B31.15 reacts with human beta Endorphin, an endogenous opiate derived from ACTH gene. ACTH (Corticotropin; human 39 aa) is synthesized by the anterior pituitary gland and stimulates the adrenal cortex; 6 hormones are derived from one ACTH gene: ACTH, lipotropin, alpha-MSH, beta-MSH, endorphin, and one other. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| B31.15 | |
| Unconjugated | |
| P01189 | |
| POMC | |
| Human beta Endorphin (full length native protein). | |
| 0.1 mg | |
| Neuroscience, Nutrient Sensing in the Brain | |
| 5443 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction