missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta Dystroglycan Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534908
This item is not returnable.
View return policy
Description
Recommended positive controls: H1299, MCF-7. Predicted reactivity: Mouse (89%), Rat (87%), Dog (89%), Cat (94%), Rabbit (89%), Rhesus Monkey (97%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Dystroglycan is a laminin binding component of the dystrophin-glycoprotein complex which provides a linkage between the subsarcolemmal cytoskeleton and the extracellular matrix. Dystroglycan 1 is a candidate gene for the site of the mutation in autosomal recessive muscular dystrophies. The dramatic reduction of dystroglycan 1 in Duchenne muscular dystrophy leads to a loss of linkage between the sarcolemma and extracellular matrix, rendering muscle fibers more susceptible to necrosis. Dystroglycan also functions as dual receptor for agrin and laminin-2 in the Schwann cell membrane. The muscle and nonmuscle isoforms of dystroglycan differ by carbohydrate moieties but not protein sequence. Alternative splicing results in multiple transcript variants all encoding the same protein.
Specifications
| beta Dystroglycan | |
| Polyclonal | |
| Unconjugated | |
| DAG1 | |
| 156DAG; A3a; AGRNR; Alpha-DG; Alpha-dystroglycan; Beta-DG; Beta-dystroglycan; D9Wsu13e; DAG; DAG1; Dag-1; DG; Dp427; Dp71; dystroglycan; dystroglycan 1; dystroglycan 1 (dystrophin-associated glycoprotein 1); dystrophin associated glycoprotein 1; Dystrophin-associated glycoprotein 1; MDDGA9; MDDGC7; MDDGC9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13138, 1605 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q14118, Q62165 | |
| DAG1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human DAG1/beta Dystroglycan. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction