missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ beta Dystroglycan Monoclonal Antibody (GT835)
GREENER_CHOICE

Product Code. 17136583 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17136583 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17136583 Supplier Invitrogen™ Supplier No. MA535931

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta Dystroglycan Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

Dystroglycan is a laminin binding component of the dystrophin-glycoprotein complex which provides a linkage between the subsarcolemmal cytoskeleton and the extracellular matrix. Dystroglycan 1 is a candidate gene for the site of the mutation in autosomal recessive muscular dystrophies. The dramatic reduction of dystroglycan 1 in Duchenne muscular dystrophy leads to a loss of linkage between the sarcolemma and extracellular matrix, rendering muscle fibers more susceptible to necrosis. Dystroglycan also functions as dual receptor for agrin and laminin-2 in the Schwann cell membrane. The muscle and nonmuscle isoforms of dystroglycan differ by carbohydrate moieties but not protein sequence. Alternative splicing results in multiple transcript variants all encoding the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Dystroglycan
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT835
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DAG1
Gene Accession No. Q14118, Q62165
Gene Alias 156DAG; A3a; AGRNR; Alpha-DG; Alpha-dystroglycan; Beta-DG; Beta-dystroglycan; D9Wsu13e; DAG; DAG1; Dag-1; DG; Dp427; Dp71; dystroglycan; dystroglycan 1; dystroglycan 1 (dystrophin-associated glycoprotein 1); dystrophin associated glycoprotein 1; Dystrophin-associated glycoprotein 1; MDDGA9; MDDGC7; MDDGC9
Gene Symbols DAG1
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human DAG1/beta Dystroglycan. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13138, 1605
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.