missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-defensin 106B Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06276PE
This item is not returnable.
View return policy
Description
Beta-defensin 106B Polyclonal antibody specifically detects Beta-defensin 106B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Beta-defensin 106B | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| BD6, BD-6, Beta-defensin 6, DEFB106, DEFB106A, DEFB106B, DEFB6, DEFB-6, Defensin, beta 106 | |
| Produced in rabbits immunized with E. coli-derived Human Beta-defensin 106B fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 503841 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction