missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta-Defensin 1 Recombinant Rabbit Monoclonal Antibody (1F5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550161
This item is not returnable.
View return policy
Description
beta-Defensin 1 Recombinant Monoclonal Antibody for Flow, ELISA
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 1, an antimicrobial peptide implicated in the resistance of epithelial surfaces to microbial colonization. This gene maps in close proximity to defensin family member, defensin, alpha 1 and has been implicated in the pathogenesis of cystic fibrosis.
Specifications
| beta-Defensin 1 | |
| Recombinant Monoclonal | |
| 0.9 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P60022 | |
| DEFB1 | |
| A synthesized peptide derived from Human DEFB1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Flow Cytometry | |
| 1F5 | |
| Unconjugated | |
| DEFB1 | |
| AW260221; BD1; BD-1; beta defensin 1; beta-defensin 1; beta-defensin-1; DEFB1; DEFB-1; DEFB101; defensin beta 1; defensin, beta 1; HBD1; rBD-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1672 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction