missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-D-Thioglucose sodium salt, 95%

Product Code. p-7043282
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15438359

Brand: Thermo Scientific Chemicals J66838.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10593-29-0 | C6H11NaO5S | 218.199 g/mol

beta-D-Thioglucose sodium salt, is used in research and to treat rheumatoid arthritis. The biological mechanism is unknown. 1-Thio-β is often used as a carrier molecule for cell uptake of markers, polymers, and nanoparticles, and potentially useful synthetic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-D-Thioglucose sodium salt, is used in research and to treat rheumatoid arthritis. The biological mechanism is unknown. 1-Thio-β is often used as a carrier molecule for cell uptake of markers, polymers, and nanoparticles, and potentially useful synthetic reagent.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10593-29-0
Molecular Formula C6H11NaO5S
Molecular Weight (g/mol) 218.199
MDL Number MFCD00061630
InChI Key VKPBZIVFRYLHPT-WNFIKIDCSA-M
Synonym sodium glucopyranosyl sulphide, glucothiose, sodium 2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-thiolate, 1-thio-beta-d-glucose sodium salt, beta-d-thioglucose, 1-, a-d-glucosylthiose, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-sodiosulfanyl oxane-3,4,5-triol, sodium 2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl sulfanide, 1-thio-, a-d-glucose sodium salt, 1-beta-d-glucosylthiose
PubChem CID 11413308
IUPAC Name sodium;(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxane-2-thiolate
SMILES C(C1C(C(C(C(O1)[S-])O)O)O)O.[Na+]

Specifications

Color White
Quantity 250 mg
Solubility Information Soluble in water
Formula Weight 218.2
Percent Purity 95%
Physical Form Powder
Chemical Name or Material beta-D-Thioglucose sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.