missing translation for 'onlineSavingsMsg'
Learn More

beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%

Product Code. 11495877
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495877 100 g 100g
11475877 5 g 5g
11485877 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495877 Supplier Thermo Scientific Alfa Aesar Supplier No. B24426.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13035-61-5 | C13H18O9 | 318.278 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13035-61-5
Molecular Formula C13H18O9
Molecular Weight (g/mol) 318.278
MDL Number MFCD00005358
InChI Key IHNHAHWGVLXCCI-FDYHWXHSSA-N
Synonym beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d
PubChem CID 83064
IUPAC Name [(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 80°C to 83°C
Quantity 100 g
Beilstein 94078
Optical Rotation −13±0.5° (c=2.4 in Chloroform)
Formula Weight 318.28
Percent Purity ≥98%
Chemical Name or Material beta-D-Ribofuranose 1,2,3,5-tetraacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.