missing translation for 'onlineSavingsMsg'
Learn More

Beta-D-Glucopyranuronic Acid Methyl Ester 2,3,4-Triacetate 1-(4-Nitrophenyl Carbonate), TRC

Product Code. 30392011
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392011 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392011 Supplier Toronto Research Chemicals Supplier No. TRCG45020010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material beta-D-Glucopyranuronic Acid Methyl Ester 2,3,4-Triacetate 1-(4-Nitrophenyl Carbonate)
CAS 228412-71-3
Molecular Formula C20H21NO14
Quantity 10 mg
Synonym [Methyl(2,3,4-tri-O-acetyl-beta-D-glucopyranosyl)uronate]-4-nitro-phenyl Carbonate
InChI Formula InChI=1S/C20H21NO14/c1-9(22)30-14-15(31-10(2)23)17(32-11(3)24)19(34-16(14)18(25)29-4)35-20(26)33-13-7-5-12(6-8-13)21(27)28/h5-8,14-17,19H,1-4H3/t14-,15-,16-,17+,19-/m0/s1
SMILES COC(=O)[C@H]1O[C@@H](OC(=O)Oc2ccc(cc2)[N+](=O)[O-])[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC Name methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(4-nitrophenoxy)carbonyloxyoxane-2-carboxylate
Molecular Weight (g/mol) 499.38
Formula Weight 499.0962
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.