missing translation for 'onlineSavingsMsg'
Learn More

Beta-D-Glucopyranosylamine, TRC

Product Code. 30397857
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397857 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397857 Supplier Toronto Research Chemicals Supplier No. TRCG41454050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material beta-D-Glucopyranosylamine
CAS 7284-37-9
Molecular Formula C6H13NO5
Quantity 50 mg
InChI Formula InChI=1S/C6H13NO5/c7-6-5(11)4(10)3(9)2(1-8)12-6/h2-6,8-11H,1,7H2/t2-,3-,4+,5-,6-/m1/s1
SMILES C(O)[C@@H]1[C@@H](O)[C@H](O)[C@@H](O)[C@H](N)O1
IUPAC Name (2R,3R,4S,5S,6R)-2-amino-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
Molecular Weight (g/mol) 179.17
Formula Weight 179.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.