missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-D-Gentiobiose, 98%

Product Code. 15458229
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
This item is not returnable. View return policy

Product Code. 15458229

Brand: Thermo Scientific Chemicals J66750.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 554-91-6 | C12H22O11

Gentiobiose was used in binding assays to determine if coelomic cytolytic factor 1 (CCF-1) binds to β-1,3-glucans to stimulate the pro-PO-activating system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gentiobiose was used in binding assays to determine if coelomic cytolytic factor 1 (CCF-1) binds to β-1,3-glucans to stimulate the pro-PO-activating system.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-91-6
Molecular Formula C12H22O11
MDL Number MFCD00198056
Synonym 6-O-(beta-D-Glucopyranosyl)-D-glucopyranose; Amygdalose

Specifications

Color White
Quantity 500 mg
Beilstein 93354
Solubility Information Slightly soluble in water.
Formula Weight 342.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material beta-D-Gentiobiose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.