missing translation for 'onlineSavingsMsg'
Learn More

Beta-D-Galactose Pentaacetate, TRC

Product Code. 30179231
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179231 25 g 25g
30179277 2.5 g 2.5g
30179331 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179231 Supplier Toronto Research Chemicals Supplier No. TRCG15576525G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4163-60-4
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.34
IUPAC Name (3,4,5,6-tetraacetyloxyoxan-2-yl)methyl acetate
SMILES CC(=O)OCC1OC(OC(=O)C)C(OC(=O)C)C(OC(=O)C)C1OC(=O)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C16H22O11/c1-7(17)22-6-12-13(23-8(2)18)14(24-9(3)19)15(25-10(4)20)16(27-12)26-11(5)21/h12-16H,6H2,1-5H3
Formula Weight 390.1162
Chemical Name or Material beta-D-Galactose Pentaacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.