missing translation for 'onlineSavingsMsg'
Learn More

beta-D-Allose, 97+%

Product Code. 11462838
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11462838

Brand: Thermo Scientific Alfa Aesar A16810.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2595-97-3 | C6H12O6 | 180.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2595-97-3
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00063268
InChI Key GZCGUPFRVQAUEE-OBOOZECYSA-N
PubChem CID 57449163
IUPAC Name (2R,3R,4R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 143°C to 146°C
Quantity 100 mg
Beilstein 1724623
Sensitivity Hygroscopic
Merck Index 14,280
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material beta-D-Allose

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.