missing translation for 'onlineSavingsMsg'
Learn More

beta-Cyclocitral, 90+%

Product Code. 11491100
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11491100

Brand: Thermo Scientific Alfa Aesar B25700.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 432-25-7 | C10H16O | 152.24 g/mol

beta-Cyclocitral is used as flavoring agent, nutrient, stabilizers, surfactants and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Cyclocitral is used as flavoring agent, nutrient, stabilizers, surfactants and emulsifiers.

Solubility
Soluble in chloroform, dichloromethane, ethyl acetate and methanol. Insoluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 432-25-7
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00079078
InChI Key MOQGCGNUWBPGTQ-UHFFFAOYSA-N
Synonym beta-cyclocitral, b-cyclocitral, 1-cyclohexene-1-carboxaldehyde, 2,6,6-trimethyl, .beta.-cyclocitral, 2,6,6-trimethyl-1-cyclohexene-1-carboxaldehyde, unii-77y0u2x29g, cyclocitral, alpha beta-cyclocitral, 2,6,6-trimethylcyclohex-1-ene-1-carbaldehyde, 1-formyl-2,6,6-trimethyl-1-cyclohexene
PubChem CID 9895
ChEBI CHEBI:53177
IUPAC Name 2,6,6-trimethylcyclohexene-1-carbaldehyde
SMILES CC1=C(C=O)C(C)(C)CCC1

Specifications

Density 0.944
Boiling Point 62°C to 63°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.498
Quantity 50 g
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,dichloromethane,ethyl acetate and methanol. Insoluble in water.
Formula Weight 152.23
Percent Purity ≥90%
Chemical Name or Material beta-Cyclocitral

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.