missing translation for 'onlineSavingsMsg'
Learn More

beta COP Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-41358CL1

Product Code. 30144940

  • £771.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

beta COP Polyclonal antibody specifically detects beta COP in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

beta COP
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
1315
Store at 4°C in the dark. Do not freeze.
IgG
Immunoprecipitation, Western Blot
CoraFluor 1
beta coat protein, Beta-coat protein, beta-cop, coatomer protein complex, subunit beta, coatomer protein complex, subunit beta 1, coatomer subunit beta, COPBFLJ57957, DKFZp761K102, FLJ10341, FLJ46444
The epitope is recognized to a region between residue 903 to 853 of human Coatomer Protein Complex, Subunit Beta 1.
0.1 mL
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.