missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-Chloromethylstyrene, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | beta-Chloromethylstyrene |
| CAS | 2687-12-9 |
| Molecular Formula | C9H9Cl |
| Quantity | 100 mg |
| Synonym | NSC 5599, Cinnamyl Chloride, 3-Phenylallyl Chloride, 3-Phenyl-2-propenyl Chloride, 3-Phenyl-2-propen-1-yl Chloride, 3-Chloroprop-1-en-1-ylbenzene, 3-Chloro-1-phenylpropene, 1-Chloro-3-phenyl-2-propene, (3-Chloro-1-propen-1-yl)benzene |
| InChI Formula | InChI=1S/C9H9Cl/c10-8-4-7-9-5-2-1-3-6-9/h1-7H,8H2/b7-4+ |
| SMILES | ClC\C=C\c1ccccc1 |
| IUPAC Name | [(E)-3-chloroprop-1-enyl]benzene |
| Molecular Weight (g/mol) | 152.62 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction