missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Catenin, p Ser552 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05441
This item is not returnable.
View return policy
Description
beta-Catenin Polyclonal antibody specifically detects beta-Catenin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| beta-Catenin | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923 | |
| A synthetic phosphorylated peptide around S552 of human beta-Catenin (NP_001091679). RTSMG | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 1499 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction